Skip to Content
Merck
All Photos(1)

Documents

05-946

Sigma-Aldrich

Anti-Trophinin Antibody, clone 3-11

culture supernatant, clone 3-11, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

3-11, monoclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TRO(7216)

Specificity

Trophinin (MAGE-D3)

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 1319-1327 (SIVGFSGGP-C) of human Trophinin (MAGE-D3)

Application

Anti-Trophinin Antibody, clone 3-11 detects level of Trophinin & has been published & validated for use in IP, WB, IH.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Routinely evaluated by immunoblot in RIPA lysates from HT-H cells

Target description

69 kDa

Physical form

Cell culture supernatant

Storage and Stability

2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The MAGE proteins: emerging roles in cell cycle progression, apoptosis, and neurogenetic disease.
Barker, Philip A and Salehi, Amir
Journal of Neuroscience Research, 67, 705-712 (2002)
Cancer/testis antigens: structural and immunobiological properties.
Kirkin, Alexei F, et al.
Cancer Investigation, 20, 222-236 (2002)
R Aoki et al.
Seminars in reproductive medicine, 18(3), 265-271 (2001-04-13)
Elucidation of the implantation mechanism in humans at the molecular level has been difficult because of methodological restrictions. Instead of using human materials during the implantation period, two human tumor cell lines that respectively mimic the biological behaviors of a
Kazuhiro Sugihara et al.
FEBS letters, 582(2), 197-202 (2007-12-15)
During embryo implantation, trophinin mediates cell adhesion by homophilic binding at the apical surfaces of trophectoderm and endometrium. Trophinin is expressed on the human endometrial epithelia in rare occasions. We developed hCG-coated agarose beads that mimic the physical and physiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service