Skip to Content
Merck
All Photos(1)

Documents

05-718

Sigma-Aldrich

Anti-PML Antibody, clone 36.1-104

clone 36.1-104, Upstate®, from mouse

Synonym(s):

Anti-MYL, Anti-PP8675, Anti-RNF71, Anti-TRIM19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

36.1-104, monoclonal

species reactivity

mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PML(5371)

General description

Promyeloctic Leukemia Gene product (PML) is a nuclear protein with growth-suppressive properties originally identified in the context of the PML-retinoic receptor alpha (RAR alpha) fusion protein of acute promyelocytic leukemia. PML localizes within distinct nuclear structures, called nuclear bodies, which are disrupted by the expression of PML-RAR alpha.

Specificity

Recognizes PML.

Immunogen

6His fusion protein corresponding to mouse PML (promyelocytic leukemia protein)

Application

Detect PML also known as Promyelocytic Leukemia Protein with Anti-PML Antibody, clone 36.1-104 (Mouse Monoclonal Antibody), that has been demonstrated to work in ICC & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

routinely evaluated by immunoblot on RIPA lysates from mouse embryo fibroblast (MEF1) cells

Target description

90-106 kDa

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide as a preservative.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
MEF1 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerdine J Stout et al.
Cancer research, 73(6), 1844-1854 (2013-01-05)
Xeroderma pigmentosum (XP), a UV-sensitivity syndrome characterized by skin hyperpigmentation, premature aging, and increased skin cancer, is caused by defects in the nucleotide excision repair (NER) pathway. XP shares phenotypical characteristics with telomere-associated diseases like Dyskeratosis congenita and mouse models
SUMO-1-dependent allosteric regulation of thymine DNA glycosylase alters subnuclear localization and CBP/p300 recruitment.
Mohan, RD; Rao, A; Gagliardi, J; Tini, M
Molecular and cellular biology null
Evgenij Evdokimov et al.
Journal of cell science, 121(Pt 24), 4106-4113 (2008-11-27)
Conjugation of the small ubiquitin-like modifier (SUMO) to target proteins regulates numerous biological processes and has been implicated in tumorigenesis and metastasis. The three SUMO isoforms in vertebrates, SUMO1 and the highly similar SUMO2 and SUMO3, can be conjugated to
SUMOylation of Tr2 orphan receptor involves Pml and fine-tunes Oct4 expression in stem cells.
Sung Wook Park, Xinli Hu, Pawan Gupta, Ya-Ping Lin, Sung Gil Ha, Li-Na Wei
Nature Structural and Molecular Biology null
Abrar Ahmed et al.
Scientific reports, 7, 44539-44539 (2017-03-21)
Promyelocytic Leukemia (PML) is a nuclear protein that forms sub-nuclear structures termed nuclear bodies associated with transcriptionally active genomic regions. PML is a tumour suppressor and regulator of cell differentiation. We demonstrate that PML promotes TNFα-induced transcriptional responses by promoting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service