Skip to Content
Merck
All Photos(2)

Key Documents

05-435

Sigma-Aldrich

Anti-Lck Antibody, clone 3A5

clone 3A5, Upstate®, from mouse

Synonym(s):

Anti-IMD22, Anti-LSK, Anti-YT16, Anti-p56lck, Anti-pp58lck

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$907.00

$907.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$907.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$907.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A5, monoclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LCK(3932)

Specificity

Recognizes Lck.

Immunogen

GST-fusion to residues 1-225 of murine Lck

Application

Research Category
Signaling
Research Sub Category
Immunological Signaling
This Anti-Lck Antibody, clone 3A5 is validated for use in IP, WB for the detection of Lck.

Quality

routinely evaluated by immunoblot on RIPA lysates from Jurkat cells

Target description

56 kDa

Linkage

Replaces: 04-372

Physical form

Format: Purified
Protein A purified
Protein A purified antibody in 100uL of 10mM PBS, pH 7.4

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The differential expression of LCK and BAFF-receptor and their role in apoptosis in human lymphomas.
Jennifer C Paterson, Sara Tedoldi, Andrew Craxton, Margaret Jones, Martin-Leo Hansmann et al.
Haematologica null
Association of SRC-related kinase Lyn with the interleukin-2 receptor and its role in maintaining constitutive phosphorylation of JAK/STAT in human T-cell leukemia virus type 1-transformed T cells.
Shuh, M; Morse, BA; Heidecker, G; Derse, D
Journal of virology null
B Omri et al.
Journal of neurochemistry, 67(4), 1360-1364 (1996-10-01)
The lck gene product, p56lck, is a member of the src-related family of protein tyrosine kinases. It is known as lymphocyte specific and involved in thymocyte development and in the immune response mediated by the T cell receptor. We report
Tiphaine Douanne et al.
Open biology, 14(2), 230456-230456 (2024-02-28)
Cytotoxic T lymphocytes (CTLs) are key effectors of the adaptive immune system that recognize and eliminate virally infected and cancerous cells. In naive CD8+ T cells, T-cell receptor (TCR) engagement drives a number of transcriptional, translational and proliferation changes over
Jing Jin et al.
Molecular and cellular biology, 36(6), 1007-1018 (2016-01-13)
The heat shock protein 90 (HSP90) and cell division cycle 37 (CDC37) chaperones are key regulators of protein kinase folding and maturation. Recent evidence suggests that thermodynamic properties of kinases, rather than primary sequences, are recognized by the chaperones. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service