Skip to Content
Merck
All Photos(1)

Key Documents

05-23-2150

Sigma-Aldrich

PACAP 38, Ovine

More active than vasoactive intestinal peptide (VIP) in stimulating adenylate cyclase (EC₅₀ = 7 nM).

Synonym(s):

PACAP 38, Ovine, Pituitary Adenylate Cyclase Activating Polypeptide, HSDGIFTDSYSRYRKQMAVKKYLAAVLGKRYKQRVKNK-NH₂

Sign Into View Organizational & Contract Pricing

Select a Size

.1 MG
$210.00

$210.00


Check Cart for Availability


Select a Size

Change View
.1 MG
$210.00

About This Item

Empirical Formula (Hill Notation):
C203H331N63O53S
CAS Number:
Molecular Weight:
4534.26
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$210.00


Check Cart for Availability

Quality Level

Assay

≥96% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

5% acetic acid: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

More active than vasoactive intestinal peptide (VIP) in stimulating adenylate cyclase (EC50 = 7 nM).

Biochem/physiol Actions

Cell permeable: no
EC50 = 7 nM against adenylate cyclase
Primary Target
Adenylate cyclase
Product does not compete with ATP.
Reversible: no

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-His-Ser-Asp-Gly-Ile-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-Gly-Lys-Arg-Tyr-Lys-Gln-Arg-Val-Lys-Asn-Lys-NH₂

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kobayashi, H., et al. 1994. Brain Res.647, 145.
Chastre, E., et al. 1991. J. Biol. Chem.266, 21239.
Le Meuth, V., et al. 1991. Am. J. Physiol. 260, G265.
Kimura, C., et al. 1990. Biochem. Biophys. Res. Commun.166, 81.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service