Skip to Content
Merck
All Photos(1)

Key Documents

05-1583

Sigma-Aldrich

Anti-PRL-2 Antibody, clone 42

clone 42, from mouse

Synonym(s):

Protein-tyrosine phosphatase 4a2, Protein-tyrosine phosphatase of regenerating liver 2, phosphatase of regenerating liver 2, protein tyrosine phosphatase IVA, protein tyrosine phosphatase IVA2, protein tyrosine phosphatase type IVA, member 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$661.00

$661.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$661.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$661.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

42, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTP4A2(8073)

General description

PRL-2 is one of three members (PRL-1, PRL-2, and PRL-3) that is part of a unique protein-tyrosine phosphatase ((PTP) family. PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. PTPs in this class contain a protein tyrosine phosphatase catalytic domain and a characteristic C-terminal prenylation motif. This PTP has been shown to primarily associate with plasmic and endosomal membrane through its C-terminal prenylation. This PTP is known to interact with the beta-subunit of Rab geranylgeranyltransferase II (beta GGT II), and thus may function as a regulator of GGT II activity. Overexpression of this gene in mammalian cells conferres a transformed phenotype and can result in a tumor growth in nude mice, suggesting a role in tumorigenesis. Alternatively spliced transcript variants that encode two distinct isoforms have been described

Specificity

This antibody recognizes PRL-2.

Immunogen

Epitope: Unknown
GST-tagged recombinant protein corresponding to mouse PRL-2.

Application

Anti-PRL-2 Antibody, clone 42 detects level of PRL-2 & has been published & validated for use in WB, IH & IP.
Immunohistochemistry Analysis:
A representative lot was used by an independent laboratory in IH.

Immunoprecipitation Analysis:
A representative lot was used by an independent laboratory in IP.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blot in HeLa cell lysate.
Western Blot Analysis: 0.5 µg/ml of this antibody detected PRL-2 on 10 µg of HeLa cell lysate.

Target description

~ 19 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noriko Uetani et al.
JCI insight, 2(13) (2017-07-07)
Magnesium (Mg2+) plays pleiotropic roles in cellular biology, and it is essentially required for all living organisms. Although previous studies demonstrated intracellular Mg2+ levels were regulated by the complex of phosphatase of regenerating liver 2 (PRL2) and Mg2+ transporter of
Xinyue Du et al.
Nature communications, 15(1), 881-881 (2024-01-30)
Excessive host immune responses contribute to severe malaria with high mortality. Here, we show that PRL2 in innate immune cells is highly related to experimental malaria disease progression, especially the development of murine severe malaria. In the absence of PRL2
Mathilde Poulet et al.
Communications biology, 3(1), 603-603 (2020-10-25)
Protein tyrosine phosphatases are essential modulators of angiogenesis and have been identified as novel therapeutic targets in cancer and anti-angiogenesis. The roles of atypical Phosphatase of Regenerative Liver (PRL) phosphatases in this context remain poorly understood. Here, we investigate the
Megan Moerdyk-Schauwecker et al.
PloS one, 9(8), e104688-e104688 (2014-08-12)
Virus particles (virions) often contain not only virus-encoded but also host-encoded proteins. Some of these host proteins are enclosed within the virion structure, while others, in the case of enveloped viruses, are embedded in the host-derived membrane. While many of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service