Skip to Content
Merck
All Photos(1)

Key Documents

05-1570

Sigma-Aldrich

Anti-PRDM1 (BLIMP-1) Antibody, clone 6D3

clone 6D3, from rat

Synonym(s):

PR domain containing 1, with ZNF domain, PR-domain zinc finger protein 1, beta-interferon gene positive-regulatory domain I binding factor, Positive regulatory domain I-binding factor 1, PRDI-binding factor-1, Beta-interferon gene positive regulatory dom

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$667.00

$667.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$667.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$667.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D3, monoclonal

species reactivity

mouse, human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRDM1(639)

General description

B-lymphocyte induced maturation protein (BLIMP-1) is a 100 kDa transcriptional repressor containing 1 DET domain and 4 C2H2-type zinc fingers. In the nucleus, BLIMP-1 is known to bind specifically with the Beta-interferon gene positive regulatory domain I (PRDI), and to interact with PRMT5. During B-cell differentiation, BLIMP-1 actively represses the transcription of the gene, c-myc, through binding its promoter and has been deemed a “master regulator” for terminal B-cell differentiation, as it is able to initiate the BCL1 cell developmental cascade.

Specificity

Recognizes BLIMP-1

Immunogen

Epitope: Recognizes epitope located between amino acids 255-395 of BLIMP-1
Recombinant protein corresponding to mouse BLIMP-1.

Application

ELISA Analysis: A representative lot was used by an independent laboratory in ELISA. (Kallies, A., et al. (2004). J Exp Med. 200(8):967-977.)

Flow Cytometry Analysis: A representative lot was used by an independent laboratory in FC. (Kallies, A., et al. (2004). J Exp Med. 200(8):967-977.)
Research Category
Epigenetics & Nuclear Function

Inflammation & Immunology
Use Anti-PRDM1 (BLIMP-1) Antibody, clone 6D3 (Rat Monoclonal Antibody) validated in WB, ELISA, IHC, FC to detect PRDM1 (BLIMP-1) also known as PR-domain zinc finger protein 1.

Quality

Evaluated by Western Blot in HL-60 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected BLIMP-1 on 10 µg of HL-60 cell lysate.

Target description

~ 95 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service