Skip to Content
Merck
All Photos(3)

Key Documents

05-1511

Sigma-Aldrich

Anti-Y14 Antibody, clone 4C4

clone 4C4, from mouse

Synonym(s):

Binder of OVCA1-1, RNA binding motif protein 8A, RNA binding motif protein 8B, RNA-binding motif protein 8A, RNA-binding protein Y14, Ribonucleoprotein RBM8A, ribonucleoprotein RBM8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$674.00

$674.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$674.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$674.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4C4, monoclonal

species reactivity

mouse, rat, human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RBM8A(9939)

General description

Y14 (RNA binding motif protein 8A or RBM8A), an RNA-binding protein, contains a conserved RNA-binding motif. Y14 is found predominantly in the nucleus, although it is also present in the cytoplasm. It is preferentially associated with mRNAs produced by splicing, including both nuclear mRNAs and newly exported cytoplasmic mRNAs. It is thought that the protein remains associated with spliced mRNAs as a tag to indicate where introns had been present, thus coupling pre- and post-mRNA splicing events. Previously, it was thought that two genes encode this protein, RBM8A and RBM8B; it is now thought that the RBM8B locus is a pseudogene. Two alternative start codons result in two forms of the protein, and this gene also uses multiple polyadenylation sites.

Specificity

This antibody recognizes Y14.

Immunogen

Epitope: Unknown
Purified human Y14.

Application

Anti-Y14 Antibody, clone 4C4 is a Mouse Monoclonal Antibody for detection of Y14 also known as Binder of OVCA1-1, RNA binding motif protein 8A, Ribonucleoprotein RBM8A & has been validated in WB, ICC & IP.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected Y14 in HeLa and NIH/3T3 cells.

Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in immunoprecipitation. Kashima, I., et al. (2006). Genes & Dev. 20:355-367.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Binding Protein (RBP)

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/ml of this antibody detected Y14 on 10 µg of HeLa cell lysate.

Target description

24 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Zhang et al.
Genetics, 200(4), 1133-1148 (2015-06-07)
In higher eukaryotes the accelerated degradation of mRNAs harboring premature termination codons is controlled by nonsense-mediated mRNA decay (NMD), exon junction complex (EJC), and nuclear cap-binding complex (CBC) factors, but the mechanistic basis for this quality-control system and the specific
Sapna Puri et al.
Nature communications, 15(1), 588-588 (2024-01-19)
Despite significant research, mechanisms underlying the failure of islet beta cells that result in type 2 diabetes (T2D) are still under investigation. Here, we report that Sox9, a transcriptional regulator of pancreas development, also functions in mature beta cells. Our

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service