Skip to Content
Merck
All Photos(3)

Documents

05-1231

Sigma-Aldrich

Anti-Progerin Antibody, clone 13A4

clone 13A4, Upstate®, from mouse

Synonym(s):

LMNA, CMD1A, LMNC, LMN1, LDP1, LFP, FPLD, FPL, EMD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13A4, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LMNA(4000)

General description

Progerin is a truncated version of lamin A protein involved in Hutchinson-Gilford progeria syndrome. Progerin is most often generated by a mutation (C1824T) in the lamin A gene, LMNA. This mutation activates a cryptic splice site and gives rise to a form of lamin A with a deletion of 50 amino acids. Lamin A constitutes a major structural component of the lamina, a scaffold of proteins found inside the nuclear membrane of a cell; progerin does not properly integrate into the lamina, which disrupts the scaffold structure and leads to significant disfigurement of the nucleus, characterized by a lobular shape. Researchers have shown that progerin activates genes that regulate stem cell differentiation via the Notch signaling pathway. Progerin, which has been linked to normal aging, is produced in healthy indivduals via "sporadic use of the cryptic splice site.
The previously assigned protein identifier Q6UYC3 has been merged into P02545. Full details can be found on the UniProt database.

Specificity

Other species not tested.
Recognizes human progerin, Mr ~70 kDa. Does not recognize Lamin A or Lamin C

Immunogen

KLH-conjugated synthetic peptide corresponding to the amino acids 604-611 of progerin (lamin A/C).

Application

Research Category
Cell Structure
This Anti-Progerin Antibody, clone 13A4 is validated for use in WB, IC, ELISA, IP for the detection of Progerin.

Quality

Routinely evaluated by western blotting on HeLa cells expressing N-terminally Flag-tagged human progerin.

Target description

70 kDa

Physical form

Format: Purified
Protein G Purified
Protein G purified immunoglobulin. Supplied at 1 mg/ml in 0.1 M Tris-Glycine (pH7.4), 150 mM NaCl, 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap.

Analysis Note

Control
HeLa cells expressing human progerin

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuela Loi et al.
Oncotarget, 7(13), 15662-15677 (2015-12-25)
Chromatin disorganization is one of the major alterations linked to prelamin A processing impairment. In this study we demonstrate that BAF is necessary to modulate prelamin A effects on chromatin structure. We show that when prelamin A and BAF cannot
Anahita Mojiri et al.
European heart journal, 42(42), 4352-4369 (2021-08-15)
Hutchinson-Gilford progeria syndrome (HGPS) is an accelerated ageing syndrome associated with premature vascular disease and death due to heart attack and stroke. In HGPS a mutation in lamin A (progerin) alters nuclear morphology and gene expression. Current therapy increases the
Asao Noda et al.
Genes and environment : the official journal of the Japanese Environmental Mutagen Society, 37, 13-13 (2015-01-01)
Progerin, the protein responsible for the Hutchinson-Gilford Progeria Syndrome (HGPS), is a partially deleted form of nuclear lamin A, and its expression has been suggested as a cause for dysfunctional nuclear membrane and premature senescence. To examine the role of
Seul-Ki Park et al.
Experimental dermatology, 26(10), 889-895 (2017-02-14)
Metformin is a popular antidiabetic biguanide, which has been considered as a candidate drug for cancer treatment and ageing prevention. Hutchinson-Gilford progeria syndrome (HGPS) is a devastating disease characterized by premature ageing and severe age-associated complications leading to death. The
Mzwanele Ngubo et al.
Aging cell, 23(7), e14150-e14150 (2024-04-05)
Hutchinson-Gilford Progeria syndrome (HGPS) is a lethal premature aging disorder caused by a de novo heterozygous mutation that leads to the accumulation of a splicing isoform of Lamin A termed progerin. Progerin expression deregulates the organization of the nuclear lamina

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service