Skip to Content
Merck
All Photos(2)

Key Documents

05-1139

Sigma-Aldrich

Anti-Focal Adhesion Kinase Antibody, clone BLAb2H7

clone BLAb2H7, from mouse

Synonym(s):

FADK 1, PTK2 protein tyrosine kinase 2, Protein-tyrosine kinase 2, focal adhesion kinase 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$613.00

$613.00


Estimated to ship on28 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1625

Request a Bulk Order

Select a Size

Change View
100 μG
$613.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$613.00


Estimated to ship on28 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1625

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BLAb2H7, monoclonal

species reactivity

human, mouse, rat, chicken

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

chicken ... Ptk2(396416)
human ... PTK2(5747)
mouse ... Ptk2(14083)
rat ... Ptk2(25614)

General description

FAK is a 125 Kd protein that is recruited as a participant in focal adhesion dynamics between cells and has a role in motility and cell survival. FAK is a highly conserved, non-receptor tyrosine kinase originally identified as a substrate for the oncogene protein tyrosine kinase v-src (Guan, J L and Shalloway, D). This cytosolic kinase has been implicated in diverse cellular roles including cell locomotion, mitogen response and cell survival. FAK is typically located at structures known as focal adhesions, these are multi-protein structures that link the extracellular matrix (ECM) to the cytoplasmic cytoskeleton. Additional components of focal adhesions include actin, filamin, vinculin, talin, paxillin and tensin (Burridge, K, et al).

Specificity

This antibody recognizes Focal Adhesion Kinase

Immunogen

Epitope: Terminal
Recombinant fragment derived from the amino terminal region of chicken FAK, amino acid residues 103 to 553.

Application

Immunohistochemistry Analysis: A previous lot detected Focal Adhesion Kinase at 1:100 dilution in mouse cortex tissue.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling
This Anti-Focal Adhesion Kinase Antibody, clone BLAb2H7 is validated for use in WB, IP for the detection of Focal Adhesion Kinase.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.
Western Blotting Analysis: 0.2 µg/mL of this antibody detected Focal Adhesion Kinase in 10 µg of HEK293 cell lysate.

Target description

~125 kDa

Linkage

Replaces: 04-591

Physical form

Format: Purified
Purified mouse monoclonal IgG1 in buffer containing 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua S McLane et al.
Biophysical journal, 109(2), 249-264 (2015-07-23)
Mechanical properties of the tumor microenvironment have emerged as key factors in tumor progression. It has been proposed that increased tissue stiffness can transform stromal fibroblasts into carcinoma-associated fibroblasts. However, it is unclear whether the three to five times increase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service