Skip to Content
Merck
All Photos(1)

Key Documents

04-889

Sigma-Aldrich

Anti-Src Antibody, clone N6L, rabbit monoclonal

culture supernatant, clone N6L, from rabbit

Synonym(s):

proto-oncogene tyrosine-protein kinase SRC, protooncogene SRC, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene

homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral onco

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$734.00

$734.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$734.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$734.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

N6L, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SRC(6714)

General description

pp60c-Src is the prototype non-receptor tyrosine kinase. Homology to Src in three regions is shared by a number of proteins: SH1 being the catalytic domain, SH2 being a phospho-tyrosine binding domain, and SH3 being a proline-binding domain. The Src SH2 domain binds to phospho-Tyr527 (phosphorylated by Csk) within the Src molecule, causing Src to adopt an inactive conformation. Mutation of Tyr527 to phenylalanine removes the SH2-binding target and results in a constitutively active kinase. A second mutation at the conserved lysine residue at the ATP-binding site abolishes kinase activity, but without Tyr527, the conformation of the protein is open, allowing interaction with endogenous activators. Consequently, this double-mutant exhibits a dominant-negative phenotype

Specificity

Recognizes Src. Crossreacts with Yes and weakly with Fyn. No crossreactivity observed with Lck or Lyn.

Immunogen

N-terminal 6His-tagged recombinant full-length human Src expressed by baculovirus in Sf21 insect cells. Purified using Ni2+/NTA agarose.

Application

Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling
This Anti-Src Antibody, clone N6L is validated for use in WB for the detection of Src.

Quality

Western Blot Analysis: A 1:500 to 1:1000 dilution of this antibody detected Src in RIPA lysates from A431 cells.

Target description

~ 60 kDa

Linkage

Replaces: 05-889

Physical form

Cultured supernantant containing 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Included positive antigen control of non-stimulated A431 lysate (Cat. No. 12-301). Add 2.5 μL of 2-mercaptoethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng-Zhe Wu et al.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology, 25(2), 159-166 (2021-02-20)
Nicotinamide adenine dinucleotide phosphate oxidases (NOXs) are the major enzymatic source of reactive oxygen species (ROS). NOX2 and NOX4 are expressed in the heart but its role in hypoxia-induced atrial natriuretic peptide (ANP) secretion is unclear. This study investigated the
Xiang Li et al.
Kardiologia polska, 79(9), 972-979 (2021-06-28)
Interleukin (IL)-18 is produced mainly in the heart and can be associated with the development of cardiac hypertrophy that leads to cardiac dysfunction. However, the effects of hypoxia on IL-18 expression and atrial natriuretic factor (ANF) secretion remain largely unknown.
Mariya S Liyasova et al.
PloS one, 14(5), e0216967-e0216967 (2019-05-24)
Many receptor tyrosine kinases (RTKs, such as EGFR, MET) are negatively regulated by ubiquitination and degradation mediated by Cbl proteins, a family of RING finger (RF) ubiquitin ligases (E3s). Loss of Cbl protein function is associated with malignant transformation driven

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service