Skip to Content
Merck
All Photos(1)

Key Documents

04-825

Sigma-Aldrich

Anti-PDGFRβ Antibody, clone 4A56, rabbit monoclonal

culture supernatant, clone 4A56, Upstate®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$743.00

$743.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$743.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$743.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

4A56, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PDGFRB(5159)

Specificity

PDGFRβ, does not cross-react with EGF receptor

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 710-718 of human PDGF Receptor β. An N-terminus cysteine was added to facilitate conjugation. This sequence corresponds to amino acids 709-717 of mouse PDGF Receptor β.

Application

Research Category
Signaling
Research Sub Category
Growth Factors & Receptors
This Anti-PDGFRβ Antibody, clone 4A56 is validated for use in IP, WB for the detection of PDGFRβ.
Westerm Blot Analysis:
A 1:2,000 dilution of this lot detected PDGFRβ in RIPA lysates from mouse NIH/3T3, 3T3/A31, and human HFF cells.

Immunoprecipitation: 4 μg of a previous lot immunoprecipitated PDGFRβ from 500 μg of 3T3/A31 RIPA lysate.

Quality

routinely evaluated by immunoblot on RIPA lysates from mouse NIH/3T3, 3T3/A31, or human HFF cells

Target description

190kDa

Linkage

Replaces: 05-825

Physical form

Cultured supernantant in 0.05% sodium azide

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap.

Analysis Note

Control
RIPA lysates from mouse NIH/3T3, 3T3/A31, or human HFF cells

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhe Wang et al.
Cancer gene therapy, 30(11), 1471-1484 (2023-08-08)
Chromosome translocations in the 5q31-33 region are associated with a range of hematologic malignancies, some of which involve the platelet-derived growth factor receptor beta (PDGFRB) gene. We report a case of acute myeloid leukemia (AML) with a mutation in the
Mohammed Hassan et al.
Cold Spring Harbor molecular case studies, 5(5) (2019-10-28)
Infantile myofibromatosis (IM) is an aggressive neoplasm composed of myofibroblast-like cells in children. Although typically localized, it can also present as multifocal disease, which represents a challenge for effective treatment. IM has previously been linked to activating somatic and germline

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service