Skip to Content
Merck
All Photos(2)

Documents

04-824

Sigma-Aldrich

Anti-Estrogen Receptor β Antibody, clone 68-4, rabbit monoclonal

culture supernatant, clone 68-4, from rabbit

Synonym(s):

Estradiol receptor, Nuclear receptor subfamily, estrogen receptor 1, estrogen receptor alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

68-4, monoclonal

species reactivity

mouse, rat, human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ESR2(2100)
mouse ... Esr2(13983)
rat ... Esr2(25149)

General description

Estrogen receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of ligand-activated transcription factors. Estrogen receptors, including ERα and ERβ, contain DNA binding and ligand binding domains and are critically involved in regulating the normal function of reproductive tissues. They are located in the nucleus, though some estrogen receptors associate with the cell surface membrane and can be rapidly activated by exposure of cells to estrogen. ERα and ERβ have been shown to be differentially activated by various ligands. Receptor-ligand interactions trigger a cascade of events, including dissociation from heat shock proteins, receptor dimerization, phosphorylation and the association of the hormone activated receptor with specific regulatory elements in target genes. Evidence suggests that ERα and ERβ may be regulated by distinct mechanisms even though they share many functional characteristics.

Specificity

Estrogen receptor β, long form, does not detect estrogen receptor alpha.
Predicted to cross-react with mouse and rat based on sequence homology.

Immunogen

Epitope: a.a.55-74(H) or 63-82 (R, M)
KLH-conjugated, synthetic peptide corresponding to amino acids 63-82 of rat and mouse estrogen receptor β and amino acids 55-74 of human estrogen receptor β.

Application

Detect Estrogen Receptor β using this Anti-Estrogen Receptor β Antibody, clone 68-4 validated for use in WB & IP.
Immunoprecipitation:
10 μL of a previous lot immunoprecipitated estrogen receptor β from 500 μg of MCF7 RIPA lysate.

Quality

Evaluated by western blot on RIPA lysates from MCF-7 cells.

Western Blot Analysis:
A 1:500 to 1:2,000 dilution of this antibody detected estrogen receptor β in RIPA lysates from MCF-7 cells.

Target description

59kDa

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

XiaoQing Wang et al.
Frontiers in endocrinology, 12, 666195-666195 (2021-09-18)
Endometriosis is an estrogen-dependent chronic disease. The abnormal proliferation and invasion of ectopic stromal cells (ESCs) are important manifestations of endometriosis, and it is necessary to find safer and more effective treatments. Extracellular vesicles (EVs) derived from human umbilical cord
Cheng Zeng et al.
Reproduction (Cambridge, England), 160(3), 481-490 (2020-06-17)
Endometriosis is an estrogen-dependent disease, and estrogen receptor 2 (ESR2) plays a critical role in the pathogenesis of ovarian endometriosis by promoting cell invasion. Yes-associated protein 1 (YAP1) plays suppressive roles in several types of tumors. However, the relationship between
F de C Gomes et al.
Andrology, 8(1), 211-220 (2019-04-24)
Nandrolone decanoate (ND) is an anabolic-androgenic steroid, and its indiscriminate use leads to subclinical alterations in the hypothalamic-pituitary-gonadal axis and androgen-dependent organs. To evaluate the effects of ND, either alone or in combination with resistance exercise (RE), on the levels
Anna Karpeta et al.
Journal of applied toxicology : JAT, 36(12), 1558-1567 (2016-03-19)
Data concerning the possible action of polybrominated diphenyl ethers (PBDEs) in hormone-dependent cancer are scarce. Some data showed that PBDEs may directly affect breast cancer cells formation and only one research showed increased proliferation of the OVCAR-3 cells, but the
Katharina Röck et al.
The Journal of biological chemistry, 287(24), 20056-20069 (2012-04-12)
Hyaluronan (HA) and versican are key components of the dermis and are responsive to ultraviolet (UV)B-induced remodeling. The aim of this study was to explore the molecular mechanisms mediating the effects of estrogen (E(2)) on HA-rich extracellular matrix during photoaging.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service