Skip to Content
Merck
All Photos(1)

Key Documents

04-811

Sigma-Aldrich

Anti-phospho-eNOS/NOS III (Thr495) Antibody, rabbit monoclonal

culture supernatant, Upstate®

Synonym(s):

NOS III, endothelial nitric oxidase synthase, endothelial nitric oxide synthase, nitric oxide synthase 3 (endothelial cell)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$816.00

$816.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$816.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$816.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

monoclonal

species reactivity

bovine

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr495)

Gene Information

mouse ... Nos3(287024)

Specificity

Predicted cross-reactivity with human, rabbit, mouse and rat
Recognizes phosphorylated eNOS/NOS III.

Immunogen

Peptide containing the sequence KK[pT]FK in which pT corresponds to phosphothreonine at residue 495 of human eNOS/NOS III

Application

This Anti-phospho-eNOS/NOS III (Thr495) Antibody is validated for use in WB for the detection of phospho-eNOS/NOS III (Thr495).
Western Blot Analysis:
A 1:2,000-1:10,000 dilution of this lot detected phosphorylated eNOS/NOS III in fetal bovine heart endothelial cell lysate.

Quality

routinely evaluated by immunoblot on fetal bovine heart endothelial cell lysate

Target description

135 kDa

Linkage

Replaces: 05-811

Physical form

Cultured supernatant in 0.05% sodium azide

Analysis Note

Control
Fetal bovine heart endothelial cell lysate

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel protective role of endogenous cardiac myocyte P2X4 receptors in heart failure.
Yang, T; Shen, JB; Yang, R; Redden, J; Dodge-Kafka, K; Grady, J; Jacobson, KA; Liang, BT
Circulation. Heart Failure null
A P S Mesquita et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 55, e11612-e11612 (2022-02-10)
Anoikis is a type of apoptosis that occurs in response to the loss of adhesion to the extracellular matrix (ECM). Anoikis resistance is a critical mechanism in cancer and contributes to tumor metastasis. Nitric oxide (NO) is frequently upregulated in
Martina Monti et al.
The Journal of pharmacology and experimental therapeutics, 351(3), 500-509 (2014-09-23)
At the cardiovascular level, nitric oxide (NO) controls smooth muscle functions, maintains vascular integrity, and exerts an antihypertensive effect. Metal-nonoates are a recently discovered class of NO donors, with NO release modulated through the complexation of the N-aminoethylpiperazine N-diazeniumdiolate ligand

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service