Skip to Content
Merck
All Photos(1)

Documents

04-557

Sigma-Aldrich

Anti-acetyl-Histone H4 Antibody, pan (Lys 5,8,12), rabbit monoclonal

culture supernatant, Upstate®

Synonym(s):

H4KAc, Histone H4 (acetyl)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

acetylation (Lys5,Lys8,Lys12)

Gene Information

human ... H4C1(8359)

Specificity

Acetylated Histone H4 on Lysine 5, 8, 12

Immunogen

Peptide corresponding to Histone H4 containing the sequence [GLG-ACK-GGA] on which Lys 12 is acetylated.

Application

Anti-acetyl-Histone H4 Antibody, pan (Lys 5, 8, 12) detects level of acetyl-Histone H4 & has been published & validated for use in WB.

Quality

Routinely evaluated by immunoblot.

Target description

11 kDa

Physical form

100 μL of rabbit monoclonal IgG hybridoma supernantent (serum free) with 0.1% NaN3.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre Bougdour et al.
The Journal of experimental medicine, 206(4), 953-966 (2009-04-08)
Plasmodium and Toxoplasma are parasites of major medical importance that belong to the Apicomplexa phylum of protozoa. These parasites transform into various stages during their life cycle and express a specific set of proteins at each stage. Although little is
Long-range transcriptional control of progesterone receptor gene expression.
Bon??y-Montoya J, Ziegler YS, Curtis CD, Montoya JA, Nardulli AM
Molecular Endocrinology null
Hira-mediated h3.3 incorporation is required for DNA replication and ribosomal RNA transcription in the mouse zygote.
Lin, CJ; Koh, FM; Wong, P; Conti, M; Ramalho-Santos, M
Developmental Cell null
Hironao Wakabayashi et al.
Epigenetics & chromatin, 10(1), 49-49 (2017-10-25)
The major human fungal pathogen Candida albicans possesses a diploid genome, but responds to growth in challenging environments by employing chromosome aneuploidy as an adaptation mechanism. For example, we have shown that C. albicans adapts to growth on the toxic
Tadashi Nakagawa et al.
iScience, 23(4), 101030-101030 (2020-04-17)
Haploinsufficiency of SETD5 is implicated in syndromic autism spectrum disorder (ASD), but the molecular mechanism underlying the pathological role of this protein has remained unclear. We have now shown that Setd5+/- mice manifest ASD-related behavioral phenotypes and that the expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service