Skip to Content
Merck
All Photos(3)

Key Documents

04-1648

Sigma-Aldrich

Goat Anti-Rabbit IgG Fab Antibody, clone RMG01

clone RMG01, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$801.00

$801.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$801.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$801.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

goat

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

clone

RMG01, monoclonal

species reactivity

rabbit

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Specificity

This antibody reacts to the Fab region of Rabbit IgG. No cross reactivity with human IgG, rat IgG, or mouse IgG.

Immunogen

Rabbit IgG

Application

Goat Anti-Rabbit IgG Fab, clone RMG01, Cat. No. 04-1648, is a highly specific goat monoclonal antibody, that targets Rabbit IgG Fab and has been tested in Western Blotting and ELISA.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling
Western Blotting Analysis (WB): 2 μg/mL from a representative lot detected human IgG in nonreduced rabbit IgG.

Quality

ELISA Analysis: The specificity and reactivity of a representative lot was confirmed by ELISA analyses.

Physical form

Format: Purified
Goat monoclonal in PBS with 1% BSA and 0.09% sodium azide.
Protein G Purified

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service