Skip to Content
Merck
All Photos(1)

Documents

04-1569

Sigma-Aldrich

Anti-RNA polymerase II subunit B1 Antibody, clone 4F8

clone 4F8, from rat

Synonym(s):

DNA-directed RNA polymerase II A, DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kd subunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, RNA polymerase II subunit B1, RNA-directed RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4F8, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

ChIP: suitable
ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POLR2B(5431)

General description

RNA polymerase II subunit B1 (RPB1) is the largest subunit of the RNA polymerase II complex. As a holoenzyme RNA polymerase II catalyzes transcription of eukaryotic DNA into RNA using the four ribonucleoside triphosphates as substrates. The RBP1 subunit, in combination with other polymerase subunits, forms a large central cleft that maintains contact between the active site of the enzyme, the DNA template, and the nascent RNA transcript. This subunit also contains a carboxy terminal domain (CTD) consisting of tandem heptapeptide repeats. In actively transcribing RNA polymerase ‘Ser-2’ and ‘Ser-5’ of the heptapeptide repeat are phosphorylated. Phosphorylation activates the RNA polymerase II beta subunit, allowing it to serve as an assembly platform for additional subunits that modulate initiation, elongation, termination and mRNA processing.

Specificity

This antibody recognizes RNA polymerase II subunit B1.

Immunogen

Epitope: Unknown
Ovalbumin-conjugated linear peptide corresponding to human RNA polymerase II subunit B1.

Application

Anti-RNA polymerase II subunit B1 Antibody, clone 4F8 is a Rat monoclonal antibody for detection of RNA polymerase II subunit B1 also known as DNA-directed RNA polymerase II A & has been validated in WB, ELISA.
Chromatin Immunoprecipitation Analysis: A representative lot was used by independent laboraotry in ChIP. (Chapman, R., et al. (2007). Science. 318(5857):1780 -1782.)
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blot in NIH/3T3 cell lysates.

Western Blot Analysis: 1 µg/ml of this antibody detected RNA polymerase II subunit B1 on 10 µg of NIH/3T3 cell lysates.

Target description

~ 220 kDa

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua E Mayfield et al.
eLife, 8 (2019-08-07)
The Positive Transcription Elongation Factor b (P-TEFb) phosphorylates Ser2 residues of the C-terminal domain (CTD) of the largest subunit (RPB1) of RNA polymerase II and is essential for the transition from transcription initiation to elongation in vivo. Surprisingly, P-TEFb exhibits
Rob D Chapman et al.
Science (New York, N.Y.), 318(5857), 1780-1782 (2007-12-15)
RNA polymerase II is distinguished by its large carboxyl-terminal repeat domain (CTD), composed of repeats of the consensus heptapeptide Tyr1-Ser2-Pro3-Thr4-Ser5-Pro6-Ser7. Differential phosphorylation of serine-2 and serine-5 at the 5' and 3' regions of genes appears to coordinate the localization of
Wenyu Wang et al.
Nature communications, 12(1), 4441-4441 (2021-07-23)
BRD4, a Bromodomain and Extraterminal (BET) protein family member, is a promising anti-cancer drug target. However, resistance to BET inhibitors targeting BRD4 is common in solid tumors. Here, we show that cancer-associated fibroblast (CAF)-activated stromal signaling, interleukin-6/8-JAK2, induces BRD4 phosphorylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service