Skip to Content
Merck
All Photos(1)

Key Documents

04-1525

Sigma-Aldrich

Anti-TAFII250 Antibody, clone 6B3

clone 6B3, from mouse

Synonym(s):

Cell cycle gene 1 protein, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa, TAF1 RNA polymerase II, TATA box binding protein-associated factor, 250kDa, TATA box binding protein (TBP)-associated factor, RNA polymerase II,

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$567.00

$567.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$567.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$567.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6B3, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TAF1(6872)

General description

TATA-binding protein associated factor 250 (TAFII250) is the largest subunit of the transcription factor TFIID. Studies have shown that TAFII250 has serine/threonine domains that can autophosphorylate and transphosphorylate the large subunit of TFIIF. More recent studies have shown that TAFII250 is also a histone acetyltransferase, suggesting that transcriptional co-activators may also have a role in targeting acetylation activity to promoters.

Specificity

This antibody recognizes TAFII250.

Immunogen

Epitope: Unknown
Human TAFII250

Application

Anti-TAFII250 Antibody, clone 6B3 is a Mouse Monoclonal Antibody for detection of TAFII250 also known as Cell cycle gene 1 protein & has been validated in WB, ICC, IHC & IP.
Immunoprecipitation
Cited by external research group using a representative lot. See Hilton, H.L., et. al (2005) Mol. Cell. Bio. 25(10):4321–4332.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/ml of this antibody detected TAFII250 on 10 µg of HeLa cell lysate.

Target description

Calculated molecular weight is 212 kDa However protein is usually observed at 250 kDa band.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service