Skip to Content
Merck
All Photos(1)

Key Documents

04-1475-I

Sigma-Aldrich

Anti-CD27 Antibody, clone RM27-3E5

clone RM27-3E5, from rat

Synonym(s):

CD27 antigen, CD27, CD27L receptor, T-cell activation antigen CD27, Tumor necrosis factor receptor superfamily member 7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$683.00

$683.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$683.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$683.00


Estimated to ship on27 April 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM27-3E5, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
neutralization: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Cd27(21940)

General description

CD27 antigen (UniProt P41272; also known as CD27, CD27L receptor, T-cell activation antigen CD27, Tumor necrosis factor receptor superfamily member 7) is encoded by the Cd27 (also known as Tnfrsf7) gene (Gene ID 21940) in murine species. CD27 is a TNF receptor superfamily member that binds to CD70 and mediates the survival, function, and differentiation of T-, B-, natural killer (NK), and plasma cells. Interaction with CD70 provides costimulation in T cells induces signaling events essential for cell proliferation, long-term maintenance of antigen-specific T cells, antiviral responses, antitumor immunity, and alloreactivity. CD27 ligation on B-cells by CD70 causes enhanced plasma cell formation and increased IgG production. CD27 is also used as a marker for memory B cells in the classification of B-cell deficiencies, including common variable immunodeficiency (CVID). CD27 is synthesized with a signal peptide (a.a. 1-23), which was removed posttranslationally to yield the mature type I transmembrane protein with an extracellular (a.a. 24-182) composed of three TNFR-Cys repeats (a.a. 26-63, 64-104, 105-141), followed by a transmembrane region (a.a. 183-203), and a cytoplasmic tail (a.a. 204-250).

Specificity

Clone RM27-3E5 enhanced anti-CD3-stimulated T-cell proliferation when immobilized in vitro and augmented OVA-specific CTL induction when administrated in vivo (Roberts, D.J., et al. (2010). J. Immunother. 33(8): 769 779; Sumi, T., et al. (2008). Immunol. Lett. 119(1-2):91-96).

Immunogen

Epitope: extracellular domain
Human IgG1 Fc-tagged recombinant mouse CD27 extracellular domain (Sumi, T., et al. (2008). Immunol. Lett. 119(1-2):91-96).

Application

Neurtralizing Analysis: A representative lot, when injected intraperitoneally following OVA-expressing B16 melanoma cells (B16-cOVA) implant, suppressed B16-cOVA lung metastasis and tumor growth among wild-type, but not Cd27-knockout, mice (Roberts, D.J., et al. (2010). J. Immunother. 33(8): 769 779).

Neurtralizing Analysis: A representative lot, when injected intraperitoneally during the induction phase of ragweed-induced experimental allergic conjunctivitis (EC), significantly affected the systemic immune responses, but not the severity of EC in mice (Sumi, T., et al. (2008). Immunol. Lett. 119(1-2):91-96).
Research Category
Inflammation & Immunology
This rat monoclonal Anti-CD27 Antibody, clone RM27-3E5, Cat. No. 04-1475-I is validated for use in Flow Cytometry and Neutralization applications for the analysis of CD27.

Quality

Evaluated by Flow Cytometry in mouse splenocytes.

Flow Cytometry Analysis: 2 µL of this antibody detected CD27-positive mouse splenocytes.

Target description

25.69/28.16 kDa (mature/pro-form) calculated.

Linkage

Replaces: 04-1475

Physical form

Format: Purified
Protein G purified.
Purified rat IgG2a in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service