Skip to Content
Merck
All Photos(1)

Documents

04-145

Sigma-Aldrich

Anti-NOD2 Antibody, clone 2D9

clone 2D9, from mouse

Synonym(s):

Caspase recruitment domain-containing protein 15, Inflammatory bowel disease protein 1, NLR family, CARD domain containing 2, NOD-like receptor C2, caspase recruitment domain family, member 15, caspase recruitment domain protein 15, nucleotide-binding ol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D9, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NOD2(64127)

General description

NOD2 (nucleotide-binding oligomerization domain containing 2), also known as the caspase recruitment domain family, member 15 (CARD15), is a member of the NOD-like Receptor (NLR) family of proteins and plays an important role in the immune system. It is an intracellular pattern recognition receptor and is linked to inflammatory diseases such as inflammatory bowel disease/Crohn′s Disease and Blau syndrome. NOD2 has two caspase recruitment domains (CARDs) and eleven leucine-rich repeats (LRRs). The protein is primarily expressed in the peripheral blood leukocytes. It plays a role in the immune response by recognizing the bacterial molecules. Recently it is has been found to interact with NLRC4 and bind to MAVS in response to ssRNA or viral RNA treatment and activate the IFN response.

Specificity

This antibody recognizes NOD2.

Immunogen

Epitope: Unknown
His-tagged recombinant protein corresponding to human NOD2.

Application

Anti-NOD2 Antibody, clone 2D9 is a Mouse Monoclonal Antibody for detection of NOD2 also known as NOD-like receptor C2 & has been validated in IHC, WB & IP.
Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling

Quality

Immunohistochemistry Analysis: A 1:25 dilution of this antibody detected NOD2 in hyperplastic colorectal carcinoma tissue.

Target description

~110 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Hyperplastic colorectal carcinoma tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subhash Mehto et al.
The EMBO journal, 41(23), e111289-e111289 (2022-10-13)
The NOD1/2-RIPK2 is a key cytosolic signaling complex that activates NF-κB pro-inflammatory response against invading pathogens. However, uncontrolled NF-κB signaling can cause tissue damage leading to chronic diseases. The mechanisms by which the NODs-RIPK2-NF-κB innate immune axis is activated and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service