Skip to Content
Merck
All Photos(1)

Key Documents

03-34-0052

Sigma-Aldrich

H-Gly-Arg-Ala-Asp-Ser-Pro-OH

Inactive control peptide for other fibronectin inhibitors.

Synonym(s):

H-Gly-Arg-Ala-Asp-Ser-Pro-OH, GRADSP

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$157.00
25 MG
$601.00

$157.00


Estimated to ship on29 May 2025



Select a Size

Change View
5 MG
$157.00
25 MG
$601.00

About This Item

Empirical Formula (Hill Notation):
C23H39N9O10
Molecular Weight:
601.61
UNSPSC Code:
12352202
NACRES:
NA.83

$157.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

5% acetic acid: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Inactive control peptide for other fibronectin inhibitors.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Inactive control peptide for other fibronectin inhibitors
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hoyt, D.G., et al. 1997. Am. J. Physiol.273, L612.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service