Skip to Content
Merck
All Photos(1)

Key Documents

03-311M

Sigma-Aldrich

Magna ChIRP Mouse XIST lncRNA Probe Set

The Magna ChIRP Mouse XIST lncRNA Probe Set contains 43 predesigned 20-mer DNA oligonucleotides tiled along and complementary to the sequence of mouse lncRNA XIST.

Sign Into View Organizational & Contract Pricing

Select a Size

12 REACTIONS
$475.00

$475.00


Estimated to ship on29 April 2025



Select a Size

Change View
12 REACTIONS
$475.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.52

$475.00


Estimated to ship on29 April 2025


Quality Level

manufacturer/tradename

Chemicon®
Magna ChIRP®

technique(s)

DNA sequencing: suitable
RNA extraction: suitable (nuclear function kit)
RNA purification: suitable (with magnetic beads)
activity assay: suitable (protein interaction)
nucleic acid detection: suitable (RNA)
protein purification: suitable (protein determination)

NCBI accession no.

General description

ChIRP (Chromatin Isolation by RNA Purification), allows recovery of specific segments of DNA and proteins using chromatin associated RNA as a target for biotinylated antisense probes. The Magna ChIRP Mouse XIST lncRNA Probe Set contains 43 predesigned 20-mer DNA oligonucleotides tiled along and complementary to the sequence of mouse lncRNA XIST. All probes are biotin-TEG-labeled at the 3-prime end. The oligonucleotides are split to two pools (even and odd) and are supplied ready for use in the ChIRP method. This probe set is fully compatible with the Maga ChIRP kits (cat. # 17-10494 and 17-10495).
Pervasive genomic transcription and the role of non-coding RNAs in this process is a rapidly evolving field that remains to be thoroughly explored in gene regulation.
Historically, chromatin immunoprecipitation (ChIP) has been used to interrogate association of proteins with genomic DNA sequences. Given that chromatin contains not only DNA and DNA-binding proteins, but also RNA-binding proteins and RNA, new methods have been developed to map the association of these RNA molecules to distinct regions of the genome. One such method, designated ChIRP (Chromatin Isolation by RNA Purification), allows recovery of specific segments of DNA and proteins using chromatin associated RNA as a target. This unique approach allows isolation of chromatin-associated RNAs (e.g. lncRNAs) within specific regions of genomic DNA sequences, as well as proteomic analysis of protein components of the lncRNA regulatory complexes. The Magna ChIRP RNA Interactome Kits (cat. # 17-10494 and 17-10495) facilitate the use of this method and has been proven compatible with this probe set.
X inactivation is an early developmental process in mammalian females that transcriptionally silences one of the pair of X chromosomes, thus providing dosage equivalence between males and females. The process is regulated by several factors, including a region of chromosome X called the X inactivation center (XIC), which encodes the lncRNA XIST (X-inactive specific transcript). Reactivation of both X chromosomes XaXa, via XIST epigenetic silencing, has been identified as a hallmark of primed to naïve stem cell conversion. Recently, using ChIRP–MS, Chu and colleagues captured 81 XIST-bound proteins in four different types of mouse embryonic stem cells during X chromosome inactivation.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
MAGNA CHIRP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service