Skip to Content
Merck
All Photos(4)

Key Documents

03-179

Sigma-Aldrich

RIPAb+ SUZ12 - RIP Validated Antibody and Primer Set

from mouse

Synonym(s):

ChET 9 protein, Chromatin precipitated E2F target 9 protein, Joined to JAZF1 protein, Suppressor of zeste 12 protein homolog, joined to JAZF1, suppressor of zeste 12 homolog (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

10 ASSAYS
$935.00

$935.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
10 ASSAYS
$935.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.52

$935.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

clone

monoclonal

species reactivity

human

manufacturer/tradename

RIPAb+
Upstate®

technique(s)

RIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... SUZ12(23512)

General description

SUZ12 is a polycomb group (PcG) protein. PcG proteins act by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development. PcG proteins are not required to initiate repression, but to maintain it during later stages of development. They probably act via the methylation of histones, rendering chromatin heritably changed in its expressibility. Component of the PRC2 complex, which methylates ′Lys-9′ and ′Lys-27′ residues of histone H3.

Specificity

SUZ12

Immunogen

Full length, recombinant human SUZ12.

Application

Immunoprecipitation from RIP lysate:
Representative lot data.
RIP lysate from HeLa cells (~2 X 10E7 cell equivalents per IP) was subjected to immunoprecipitation using either 5 µg of a normal mouse IgG (Cat. #CS200621), or 5 µg of Anti-SUZ12 antibody (Cat. # CS203201).
20 percent of the precipitated proteins (lane 1: mouse IgG, lane 2: SUZ12) were resolved by electrophoresis, transferred to nitrocellulose and probed with anti-SUZ12 antibody (Cat. # CS203201, 1:1000). Proteins were visualized using One-Step IP-Western kit (GenScript Cat. # L00232)
Arrow indicates SUZ12 (Figure 2).

Western Blot Analysis:
Representative lot data.
Lysates from HeLa cells were resolved by electrophoresis, transferred to PVDF and probed with anti-SUZ12 (1:1,000 dilution).
Proteins were visualized using goat anti-mouse secondary antibody conjugated to HRP and chemiluminescence detection.
Arrow SUZ12 (~95 kDa) (Figure 3).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This RIPAb+ SUZ12 -RIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

10 assays per set. Recommended use: ~5 μg of antibody per RIP (dependent upon biological context).

Quality

RNA Binding Protein Immunoprecipitation:
RIP Lysate prepared from HeLa cells (2 X 10E7 cell equivalents per IP) were subjected to immunoprecipitation using either 5 µg of a normal mouse IgG , or 5 µg of Anti-SUZ12 antibody and the Magna RIP® RNA-Binding Protein Immunoprecipitation Kit (Cat. # 17-700).
Successful immunoprecipitation of SUZ12-associated RNA was verified by qPCR using RIP Primers lincSFPQ, (Figure 1)
Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.

Target description

95 kDa

Physical form

Anti-SUZ12 (Mouse Monoclonal) -
One vial containing 50 μg of protein A purified antibody in PBS containing 0.1% azide, before the addition of 30% glycerol. Store at -20°C.

Normal Mouse IgG -
One vial containing 125 µg of purified mouse IgG in 125 µL of storage buffer containing 0.1% sodium azide. Store at -20°C.

RIP Primers, lincSFPQ -
One vial containing 75 μL of 5 μM of each primer specific for the human lincRNA SFPQ. Store at -20°C.
FOR: TCG TAC TGT TAG GCC CTT GG
REV: AAC CTT GCA TGA AGA GCA CC
Format: Purified
Protein A purified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Includes negative control normal mouse IgG antibody and control primers specific for the cDNA of human lincRNA SFPQ.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MAGNA RIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felipe C Beckedorff et al.
PLoS genetics, 9(8), e1003705-e1003705 (2013-08-31)
The down-regulation of the tumor-suppressor gene RASSF1A has been shown to increase cell proliferation in several tumors. RASSF1A expression is regulated through epigenetic events involving the polycomb repressive complex 2 (PRC2); however, the molecular mechanisms modulating the recruitment of this
Jing Wang et al.
Cancer medicine, 8(6), 3026-3035 (2019-04-25)
Ovarian cancer is one of the most common female reproductive system malignancies worldwide. Recently, the aberrant long noncoding RNAs (lncRNAs) expression has been identified in multiple cancers. Emerging evidence has highlighted the critical roles of lncRNAs in carcinogenesis and tumor
Fuzhen Qi et al.
Journal of hematology & oncology, 10(1), 48-48 (2017-02-18)
Long noncoding RNAs (lncRNAs) have emerged as important regulators of tumorigenesis and cancer progression. Recently, the lncRNA AGAP2-AS1 was identified as an oncogenic lncRNA in human non-small cell lung cancer (NSCLC) and its elevated expression was linked to NSCLC development

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service