Skip to Content
Merck
All Photos(1)

Key Documents

03-0213-00

Millipore

SMC® Human IFNλ1 High Sensitivity Immunoassay Kit

SMC® advanced ultrasensitive immunoassay technology

Synonym(s):

SMC® Human IFN Lambda 1 kit, Interferon Lambda 1 high sensitivity kit, IFN ultra-sensitive kit, Interferon Lambda 1 assay, SMC® IL-29 high sensitivity assay, IL-29 high sensitivity assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
Pricing and availability is not currently available.

description

100µL per well of 1:2 diluted serum and plasma is required

Quality Level

product line

SMC®

species reactivity

human

assay range

intra-assay cv: 5%
sensitivity: 0.26 pg/mL
(LLOQ)

standard curve range: 0.13-200 pg/mL
inter-assay cv: 9%

technique(s)

single molecule counting: suitable

input

plasma (K2EDTA)
serum

shipped in

wet ice

storage temp.

2-8°C

General description

Interferon lambda 1 (IFN-λ1), also known as IL-29, is a type of cytokine belonging to the Type III interferon family, which also includes IL-28A and IL-28B. These cytokines are recognized for their role in the immune response against viral infections and are structurally similar to members of the IL-10 family. IL-29 is primarily produced by dendritic cells and is involved in the modulation of antiviral activity, acting similarly to type I interferons, though it interacts with a distinct receptor complex. This cytokine is crucial for its antiviral properties, immune regulation, and potential therapeutic implications in managing viral infections and certain autoimmune disorders. The specific signaling through its receptor can activate various pathways leading to the induction of antiviral state and regulation of immune responses.

Application

For ultrasensitive detection of IFNλ1 (IL-29) in human serum and plasma

Features and Benefits

This SMC® kit features

  • Accurate and reproducible performance on the state-of-the-art SMCxPRO® ultrasensitive immunoassay system.
  • Compatiblity with automated liquid handling

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Explore ultrasensitive immunoassay technology with SMC® technology, including its biomarker detection platform, advantages of high sensitivity assays, and more.

Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service