Skip to Content
Merck
All Photos(1)

Key Documents

KEY465196098

Key Organics / BIONET

[2-(4-methoxyphenyl)cyclopropyl](phenyl)methanone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
252.31
MDL number:

Iupac Name

[2-(4-methoxyphenyl)cyclopropyl](phenyl)methanone

Inchi Code

1S/C17H16O2/c1-19-14-9-7-12(8-10-14)15-11-16(15)17(18)13-5-3-2-4-6-13/h2-10,15-16H,11H2,1H3

InChI key

YJPKQLQORKPVBN-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Liquid

Product Link

https://www.keyorganics.net/sku/4N-017

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/4N-017


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library