Skip to Content
Merck
All Photos(1)

Documents

JWPH3249C76E

J & W Pharmlab LLC

2,4-Dioxo-1,3-diaza-spiro[4.5]decane-8-carboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
268.31
MDL number:

Iupac Name

tert-butyl 2,4-dioxo-1,3-diazaspiro[4.5]decane-8-carboxylate

Inchi Code

1S/C13H20N2O4/c1-12(2,3)19-9(16)8-4-6-13(7-5-8)10(17)14-11(18)15-13/h8H,4-7H2,1-3H3,(H2,14,15,17,18)

InChI key

NTANNQQRKPIVQJ-UHFFFAOYSA-N

Purity

98%

Country of Origin

US

Physical Form

White Solid

Product Link

https://www.jwpharmlab.com/home/product/15R0510

MSDS

https://www.jwpharmlab.com/home/product/15R0510


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service