ENA429361254
Enamine
4-chloro-7-hydroxy-1,3-dihydro-2-benzofuran-1-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
158-159
Iupac Name
4-chloro-7-hydroxyisobenzofuran-1(3H)-one
Inchi Code
1S/C8H5ClO3/c9-5-1-2-6(10)7-4(5)3-12-8(7)11/h1-2,10H,3H2
InChI key
VZYMMKHAIQYSNS-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-192163
Melting Point
158-159
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-192163
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.