ENA311759678
Enamine
1-chloro-4-(prop-2-yn-1-yloxy)benzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-chloro-4-(2-propynyloxy)benzene
Inchi Code
1S/C9H7ClO/c1-2-7-11-9-5-3-8(10)4-6-9/h1,3-6H,7H2
InChI key
USRMDNZIKVMDKH-UHFFFAOYSA-N
storage temp.
4
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
liquid
Product Link
https://www.enaminestore.com/catalog/EN300-70771
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-70771
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service