Skip to Content
Merck
All Photos(1)

Documents

CIAH987EDE08

ChemScene LLC (Preferred Partner)

(Tetrahydro-2H-pyran-4-yl)hydrazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
116.16
MDL number:

bp

234.494C at 760 mmHg

Iupac Name

1-tetrahydro-2H-pyran-4-ylhydrazine

Inchi Code

1S/C5H12N2O/c6-7-5-1-3-8-4-2-5/h5,7H,1-4,6H2

InChI key

CMXVPHSHKFQSHM-UHFFFAOYSA-N

storage temp.

-20C, stored under nitrogen

Purity

98%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/116312-69-7.html

Flash Point

95.6C

Boiling point

234.494C at 760 mmHg

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-B0289.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service