Skip to Content
Merck
All Photos(1)

Documents

C12H316CF13E

Chemcia Scientific, LLC

7-Bromo-6-iodo-3H-quinazolin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
350.94
MDL number:

Iupac Name

7-bromo-6-iodoquinazolin-4-ol

Inchi Code

1S/C8H4BrIN2O/c9-5-2-7-4(1-6(5)10)8(13)12-3-11-7/h1-3H,(H,11,12,13)

InChI key

AZYVZRLMXLBRQV-UHFFFAOYSA-N

storage temp.

0-5

Purity

95%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Brown Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=6831

MSDS

http://www.chemcia.com/product_fl02.asp?id=6831


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service