T-041
Topiramate-D12 solution
100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
About This Item
Recommended Products
grade
certified reference material
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
clinical testing
format
single component solution
storage temp.
−20°C
SMILES string
[2H]C([2H])([2H])C1(O[C@H]2CO[C@@]3(COS(N)(=O)=O)OC(O[C@H]3[C@H]2O1)(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C12H21NO8S/c1-10(2)18-7-5-16-12(6-17-22(13,14)15)9(8(7)19-10)20-11(3,4)21-12/h7-9H,5-6H2,1-4H3,(H2,13,14,15)/t7?,8?,9-,12-/m0/s1/i1D3,2D3,3D3,4D3
InChI key
KJADKKWYZYXHBB-FTZSVORRSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Quantitative determination of topiramate in human plasma: A method employing liquid chromatography tandem mass spectrometry was developed to quantitatively determine topiramate in human plasma, highlighting the precision and applicability of using advanced analytical techniques for therapeutic drug monitoring in clinical settings (Popov et al., 2013).
- Dried blood spots technique for topiramate: The development and validation of a dried blood spots technique for quantitative determination of topiramate using liquid chromatography-tandem mass spectrometry was conducted. This method offers a minimally invasive sample collection option that could improve therapeutic drug monitoring for patients on topiramate (Popov et al., 2013).
- Therapeutic drug monitoring by LC-MS/MS: A study described a robust method for the therapeutic drug monitoring of topiramate using liquid chromatography-tandem mass spectrometry (LC-MS/MS), providing essential data on drug stability and concentration levels necessary for effective patient management (Matar, 2010).
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service