T-027
cis-Tramadol hydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
About This Item
grade
certified reference material
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1.0 mg/mL in methanol (as free base)
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
Cl.COc1cccc(c1)[C@@]2(O)CCCC[C@@H]2CN(C)C
InChI
1S/C16H25NO2.ClH/c1-17(2)12-14-7-4-5-10-16(14,18)13-8-6-9-15(11-13)19-3;/h6,8-9,11,14,18H,4-5,7,10,12H2,1-3H3;1H/t14-,16+;/m1./s1
InChI key
PPKXEPBICJTCRU-XMZRARIVSA-N
General description
Application
- Enantiomeric Analysis in Pharmacokinetics: A study detailed the quantitation of tramadol and its metabolites, including the enantiomers, in human whole blood using liquid chromatography-mass spectrometry (LC-MS/MS). This research is vital for understanding the pharmacokinetics of cis-Tramadol, supporting its application in pain management and enhancing the accuracy of pharmaceutical research in analgesic efficacy and safety (Haage et al., 2016).
- Psychoactive Drug Detection: Another study developed a multicomponent LC-MS/MS screening method for the detection of new psychoactive substances, including tramadol, in urine. This method is crucial for forensic and clinical toxicology, providing insights into the abuse potential of substances like cis-Tramadol and aiding in the regulatory monitoring of controlled substances (Al-Saffar et al., 2013).
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service