Skip to Content
Merck
All Photos(1)

Key Documents

S-045

Supelco

JWH-250 5-Hydroxypentyl metabolite solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

JWH-250 5-Hydroxypentyl metabolite solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25NO3
CAS Number:
Molecular Weight:
351.44
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

COc1ccccc1CC(=O)c2cn(CCCCCO)c3ccccc23

InChI

1S/C22H25NO3/c1-26-22-12-6-3-9-17(22)15-21(25)19-16-23(13-7-2-8-14-24)20-11-5-4-10-18(19)20/h3-6,9-12,16,24H,2,7-8,13-15H2,1H3

InChI key

BEAGHVHOGRTTHK-UHFFFAOYSA-N

General description

A primary urinary metabolite of JWH-250, a synthetic cannabinoid found in certain herbal blends of K2 or Spice. This monohydroxy analog is also referred to as the omega-hydroxy metabolite of JWH-250.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service