Skip to Content
Merck
All Photos(1)

Documents

S-023

Supelco

Sirolimus-D3 solution

100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H76D3NO13
CAS Number:
Molecular Weight:
917.19
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

application(s)

clinical testing

format

single component solution

shipped in

dry ice

storage temp.

−70°C

General description

A stable-labeled internal standard for the immunosuppressant sirolimus, an immunosuppressant and narrow therapeutic index drug used to prevent rejection in organ transplantation. This ertified calibration material is suitable for critical quantitative LC-MS/MS applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the drug′s narrow therapeutic range.

Application



  • Development of generic metabolic Raman calibration models: This study applied solution titration in aqueous phases and data augmentation techniques to develop robust Raman spectroscopy calibration models, potentially useful for in-line monitoring of bioprocesses including those involving Sirolimus-D₃ (Zhang et al., 2024).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service