Skip to Content
Merck
All Photos(1)

Key Documents

S-012

Supelco

Salvinorin A solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28O8
CAS Number:
Molecular Weight:
432.46
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

COC(=O)[C@@H]1C[C@H](OC(C)=O)C(=O)[C@H]2[C@@]1(C)CC[C@H]3C(=O)O[C@@H](C[C@]23C)c4ccoc4

InChI

1S/C23H28O8/c1-12(24)30-16-9-15(20(26)28-4)22(2)7-5-14-21(27)31-17(13-6-8-29-11-13)10-23(14,3)19(22)18(16)25/h6,8,11,14-17,19H,5,7,9-10H2,1-4H3/t14-,15-,16-,17-,19-,22-,23-/m0/s1

InChI key

OBSYBRPAKCASQB-AGQYDFLVSA-N

General description

Salvinorin A, listed on the DEA′s Drugs and Chemicals of Concern website, is the main psychotropic agent of the hallucinogenic drug Salvia also known as Sally-D, Magic Mint, and Seer′s Sage. Salvinorin A is considered the most potent naturally occurring hallucinogen.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo R Butelman et al.
The Journal of pharmacology and experimental therapeutics, 341(3), 802-808 (2012-03-22)
Active blood-brain barrier mechanisms, such as the major efflux transporter P-glycoprotein (mdr1), modulate the in vivo/central nervous system (CNS) effects of many pharmacological agents, whether they are used for nonmedical reasons or in pharmacotherapy. The powerful, widely available hallucinogen salvinorin
Zhenhong Wang et al.
PloS one, 7(7), e41724-e41724 (2012-08-23)
Cerebral hypoxia/ischemia (HI) is not uncommon during the perinatal period. If occurring, it can result in severe neurologic disabilities that persist throughout life. Salvinorin A, a non-opioid Kappa opioid receptors (KOR) selective agonist, has the potential to address this devastating
Mitchell T Harden et al.
Behavioural pharmacology, 23(7), 710-715 (2012-08-29)
Salvinorin A (SalvA), the hallucinogenic derivative of the plant Salvia divinorum, is a selective κ-opioid receptor agonist that may also have antidepressant properties. Chronic mild stress (CMS) was applied to male and female Long-Evans rats to model anhedonia common in
Joanna Listos et al.
Pharmacological reports : PR, 63(6), 1305-1309 (2012-02-24)
The hallucinogenic plant Salvia divinorum (i.e., "magic mint") is a member of the Sage family that has been historically used for divination and shamanism by the Mazatecs. Today, S. divinorum has become increasingly popular as a recreational drug for its
Melissa A Bodnar Willard et al.
Analytical and bioanalytical chemistry, 402(2), 833-842 (2011-11-01)
Salvia divinorum is a hallucinogenic herb that is internationally regulated. In this study, salvinorin A, the active compound in S. divinorum, was extracted from S. divinorum plant leaves using a 5-min extraction with dichloromethane. Four additional Salvia species (Salvia officinalis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service