Skip to Content
Merck
All Photos(1)

Key Documents

O-025

Supelco

Oxcarbazepine solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

NC(=O)N1c2ccccc2CC(=O)c3ccccc13

InChI

1S/C15H12N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8H,9H2,(H2,16,19)

InChI key

CTRLABGOLIVAIY-UHFFFAOYSA-N

General description

Oxcarbazepine, marketed under the trade name Trileptal®, is an anticonvulsant developed and prescribed for treatment of epilepsy. In recent years, Oxcarbazepine has shown efficacy in treatment of mood disorders. This certified solution standard is suitable as starting material for the preparation of calibrators and controls in oxcarbazepine testing by GC/MS or LC-MS/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Trileptal is a registered trademark of Novartis Corporation

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Régis Bouquié et al.
Therapie, 65(1), 61-65 (2010-03-09)
Oxcarbazepine is an analogue of carbamazepine, used for the treatment of partial seizure with or without secondary generalization. The two forms R and S of the mono-hydroxylated derivatives (MHD) are responsible for most of the anti-convulsant activity and it is
Ekaterine Popova et al.
Expert review of neurotherapeutics, 7(6), 617-626 (2007-06-15)
Oxcarbazepine is an antiepileptic drug that has been approved by the US FDA and is indicated for use as monotherapy or adjunctive therapy in the treatment of partial seizures in adults and children aged over 4 years. The aim of
A Vasudev et al.
The Cochrane database of systematic reviews, (1)(1), CD005171-CD005171 (2008-02-07)
Some studies have suggested that oxcarbazepine has a role in preventing episode recurrence in bipolar affective disorder. This review attempted to investigate the existing evidence from randomised controlled trials for its use in the maintenance treatment of this illness. To
Barbara R Sommer et al.
Expert opinion on drug safety, 6(2), 133-145 (2007-03-21)
Few controlled studies are available to guide the clinician in treating potentially assaultive elderly individuals with psychiatric disorders. Safety concerns limit the use of benzodiazepines and antipsychotic medications in the elderly individual, making anticonvulsants an attractive alternative. This paper reviews
Gaetano Zaccara et al.
Seizure, 22(7), 528-536 (2013-04-30)
Analysis of overall tolerability and neurological adverse effects (AEs) of eslicarbazepine acetate (ESL), lacosamide (LCM) and oxcarbazepine (OXC) from double-blind, placebo-controlled trials. Indirect comparisons of patients withdrawing because of AEs, and the incidence of some vestibulocerebellar AEs between these three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service