Skip to Content
Merck
All Photos(1)

Key Documents

M-919

Supelco

Mianserin hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21ClN2
CAS Number:
Molecular Weight:
300.83
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CN1CCN2C(C1)c3ccccc3Cc4ccccc24

InChI

1S/C18H20N2.ClH/c1-19-10-11-20-17-9-5-3-7-15(17)12-14-6-2-4-8-16(14)18(20)13-19;/h2-9,18H,10-13H2,1H3;1H

InChI key

YNPFMWCWRVTGKJ-UHFFFAOYSA-N

General description

Mianserin is a tetracyclic antidepressant prescribed for the treatment of depression and is sold under the trade names Lumin and Tolvon. This certified solution standard is applicable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, and forensic analysis. This tetracyclic antidepressant functions as a noradrenergic and a specific serotonergic antidepressant with anxiolytic, hypnotic, antiemetic, orexigenic, and antihistamine effects.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seda G Sagdinc et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 104, 222-234 (2012-12-26)
Mirtazapine (±)-1,2,3,4,10,14b-hexahydro-2-methylpyrazino(2,1-a)pyrido(2,3-c)(2)benzazepine is a compound with antidepressant therapeutic effects. It is the 6-aza derivative of the tetracyclic antidepressant mianserin (±)-2-methyl-1,2,3,4,10,14b-hexahydrodibenzo[c,f]pyrazino[1,2-a]azepine. The FT-IR and FT-Raman spectra of mirtazapine have been recorded in 4000-400 cm(-1) and 3500-10 cm(-1), respectively. The optimized geometry
Mustafa Gulec et al.
Psychiatry and clinical neurosciences, 67(1), 50-58 (2013-01-03)
Cisplatin chemotherapy is associated with neurotoxicity, and oxidative stress might play an important role in the pathogenesis. Mirtazapine may be a preventative agent via its less-known antioxidant properties. The aim of this study was to examine the potential chemoprotective effects
Marij Zuidersma et al.
Journal of psychosomatic research, 74(1), 25-30 (2013-01-01)
Evaluating the effects of implementing an antidepressant treatment strategy in depressed myocardial infarction (MI)-patients on long-term cardiovascular outcomes and all-cause mortality. MI-patients were evaluated for the presence of a diagnosis of post-MI depression at 3, 6, 9 and 12months after
Clinical practice. Chronic pruritus.
Gil Yosipovitch et al.
The New England journal of medicine, 368(17), 1625-1634 (2013-04-26)
Eveline Jaquenoud Sirot et al.
Journal of clinical psychopharmacology, 32(5), 622-629 (2012-08-29)
Pharmacogenetic tests and therapeutic drug monitoring may considerably improve the pharmacotherapy of depression. The aim of this study was to evaluate the relationship between the efficacy of mirtazapine (MIR) and the steady-state plasma concentrations of its enantiomers and metabolites in

Articles

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service