Skip to Content
Merck
All Photos(1)

Key Documents

M-259

Supelco

(±)-Methamphetamine-13C6 hydrochloride solution

new

100 μg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C413C6H15N · HCl
CAS Number:
Molecular Weight:
191.65
MDL number:
UNSPSC Code:
12352200
Product M-259 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

format

single component solution

storage temp.

-10 to -25°C

General description

Methamphetamine is a psychostimulant drug of the phenethylamine and amphetamine classes known by the street names "meth," "ice," and "glass". This internal standard is suitable for quantitation of methamphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, sports testing, forensic analysis, or isotope dilution methods. The dextrorotatory isomer of methamphetamine is a pharmaceutical drug sold under the trade name Desoxyn® for treatment of ADHD and obesity.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Desoxyn is a registered trademark of Abbott Laboratories Corp.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service