Skip to Content
Merck
All Photos(1)

Documents

M-172

Supelco

3-Methoxytyramine-D4 HCl

100 μg/mL in methanol (as free base), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9D4NO2 · HCl
CAS Number:
Molecular Weight:
207.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC1=C(OC)C=C(C([2H])([2H])C([2H])([2H])N)C=C1.[H]Cl

InChI

1S/C9H13NO2.ClH/c1-12-9-6-7(4-5-10)2-3-8(9)11;/h2-3,6,11H,4-5,10H2,1H3;1H/i4D2,5D2;

InChI key

AWRIOTVUTPLWLF-HGFPCDIYSA-N

General description

A stable-labeled internal standard suitable for LC-MS/MS monitoring of 3-methoxytyramine levels in plasma or urine for diagnostic testing, endocrinology, or clinical chemistry. 3-Methoxytyramine is a metabolite of dopamine, a catecholamine which acts as a neurotransmitter for dopamine receptors and a neurohormone for regulation of numerous brain functions. 3-Methoxytyramine levels can serve as a biomarker for the diagnosis of neuroendocrine tumors such as pheochromocytoma or paraganglioma.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service