Skip to Content
Merck
All Photos(1)

Documents

M-159

Supelco

(±)-4-Methyl-N-ethyl-norephedrine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19NO · HCl
Molecular Weight:
229.75
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

format

single component solution

storage temp.

−20°C

SMILES string

CC(C(O)C1=CC=C(C)C=C1)NCC.Cl

InChI

1S/C12H19NO.ClH/c1-4-13-10(3)12(14)11-7-5-9(2)6-8-11;/h5-8,10,12-14H,4H2,1-3H3;1H

InChI key

OILWAWJXJPGPCJ-UHFFFAOYSA-N

General description

(±)-4-Methyl-N-ethyl-norephedrine HCl is a major urinary metabolite of 4-Methylethcathinone. 4-Methylethcathinone, also known as 4-MEC, is a synthetic cathinone found in numerous bath salt blends including NRG-1 and NRG-2. This certified Snap-N-Spike® solution is suitable for LC/MS or GC/MS 4-MEC testing methods in forensic analysis, clinical toxicology, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service