Skip to Content
Merck
All Photos(1)

Key Documents

H-172

Supelco

4-Hydroxymidazolam solution

1 mg/mL (90:10 Acetonitrile:DMSO), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

4-Hydroxymidazolam solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13ClFN3O
CAS Number:
Molecular Weight:
341.77
MDL number:
UNSPSC Code:
12161501
NACRES:
NA.24
Product H-172 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (90:10 Acetonitrile:DMSO)

format

single component solution

storage temp.

-10 to -25°C

SMILES string

Fc1c(cccc1)C2=NC(c3[n](c(nc3)C)c4c2cc(cc4)Cl)O

InChI

1S/C18H13ClFN3O/c1-10-21-9-16-18(24)22-17(12-4-2-3-5-14(12)20)13-8-11(19)6-7-15(13)23(10)16/h2-9,18,24H,1H3

InChI key

ZYISITHKPKHPKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Snap-N-Spike®/Snap-N-Shoot®
This Certified Spiking Solution® is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing. 4-Hydroxymidazolam is the minor hydroxylated metabolite of midazolam (MDZ). The product contributes in the pharmacological impact of MDZ.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service