Skip to Content
Merck
All Photos(1)

Documents

H-037

Supelco

Heroin-D9 solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14NO5D9
CAS Number:
Molecular Weight:
378.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])N1CC[C@]23[C@H]4Oc5c(OC(=O)C([2H])([2H])[2H])ccc(C[C@@H]1[C@@H]2C=C[C@@H]4OC(=O)C([2H])([2H])[2H])c35

InChI

1S/C21H23NO5/c1-11(23)25-16-6-4-13-10-15-14-5-7-17(26-12(2)24)20-21(14,8-9-22(15)3)18(13)19(16)27-20/h4-7,14-15,17,20H,8-10H2,1-3H3/t14-,15+,17-,20-,21-/m0/s1/i1D3,2D3,3D3

InChI key

GVGLGOZIDCSQPN-OYCCCKEBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard with suitable uses in LC/MS and GC/MS application for quantitation of heroin levels in clinical toxicology, forensic analysis and urine drug testing applications. Heroin, also known as diacetylmorphine, is an opiate analgesic most commonly used as an illcit recreational drug.

Application


  • Controlled heroin-maintenance research: A study conducted with participants in a controlled heroin-maintenance program analyzed hair samples to establish dose-concentration relationships, utilizing Heroin-D₉ as a stable isotopic label to trace heroin metabolites, demonstrating its utility in pharmacokinetic research (Kintz et al., 1998).

  • Sweat testing for heroin maintenance: Heroin-D₉ was used in sweat testing to monitor heroin and its metabolites within a heroin maintenance program, highlighting its effectiveness as an internal standard for analytical research in clinical settings (Kintz et al., 1997).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service