Skip to Content
Merck
All Photos(1)

Key Documents

H-003S

Supelco

Hydrocodone solution

1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Hydrocodone solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO3
CAS Number:
Molecular Weight:
299.36
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

format

single component solution

storage temp.

-10 to -25°C

SMILES string

N1([C@H]2[C@H]3[C@@]4([C@@H](Oc5c4c(ccc5OC)C2)C(=O)CC3)CC1)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O.O

InChI

1S/C18H21NO3.C4H6O6.H2O/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19;5-1(3(7)8)2(6)4(9)10;/h3,6,11-12,17H,4-5,7-9H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10);1H2/t11-,12+,17-,18-;1-,2-;/m01./s1

InChI key

ZICRJLQTAUVSDU-NSJZFRMTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrocodone is a semi-synthetic opioid derived from codeine or thebaine, two naturally-occurring opiates. The drug, also known as dihydrocodeinone, is used as an analgesic to treat pain and as an antitussive for coughing or related conditions. This Certified Spiking Solution® is suitable as a starting material for calibrators or controls in GC/MS or LC/MS testing applications from pharmaceutical research and pain prescription monitoring to clinical toxicology and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service