Skip to Content
Merck
All Photos(1)

Key Documents

ERT-113S

Supelco

Tetramethylenedisulfotetramine solution

100 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N4O4S2
CAS Number:
Molecular Weight:
240.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

application(s)

agriculture
environmental

format

single component solution

storage temp.

−20°C

SMILES string

O=S1(=O)N2CN3CN1CN(C2)S3(=O)=O

InChI

1S/C4H8N4O4S2/c9-13(10)5-1-6-3-8(13)4-7(2-5)14(6,11)12/h1-4H2

InChI key

AGGKEGLBGGJEBZ-UHFFFAOYSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian P Mayer et al.
Magnetic resonance in chemistry : MRC, 50(3), 229-235 (2012-03-03)
The binding stoichiometry, strength and structure of inclusion complexes formed between the neurotoxin tetramethylenedisulfotetramine (TETS) and both native and modified cyclodextrins (CyDs) were investigated using nuclear magnetic resonance (NMR) spectroscopy. Of all six examined cases, native β-cyclodextrin (β-CyD) and its
Utcharee Intusoma et al.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 92(10), 1393-1395 (2009-10-23)
A cluster of patients with tetramethylenedisulfotetramine (TETS) intoxication was reported in Thailand. Two siblings, a-six-month-old boy and a-four-year-old girl, and their domestic dogs presented with status epilepticus within 10 minutes after ingesting milk prepared from the same tin container of
Elizabeth Hamelin et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(27), 2541-2547 (2010-04-16)
Tetramethylene disulfotetramine (tetramine) is a rodenticide associated with numerous poisonings was extracted and quantified in human urine using both gas chromatography/mass spectrometry (GC/MS) and GC/tandem mass spectrometry (MS/MS). 1200 μL samples were prepared using a (13)C(4)-labeled internal standard, a 96-well
Dorota Zolkowska et al.
The Journal of pharmacology and experimental therapeutics, 341(2), 435-446 (2012-02-14)
Tetramethylenedisulfotetramine (tetramine; TETS) is a potent convulsant poison that is considered to be a chemical threat agent. To provide a basis for the investigation of antidotes for TETS-induced seizures, we characterized the convulsant activity of TETS in mice and rats
Li Zhao et al.
ChemSusChem, 3(7), 840-845 (2010-06-25)
The synthesis of carbonaceous materials with a high surface density of amino functions for CO(2) sorption and sequestration is reported. The amino-rich carbonaceous materials are characterized by elemental analysis, N(2) sorption, scanning and transmission electron microscopy, zeta potential, TGA and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service