Skip to Content
Merck
All Photos(1)

Key Documents

E-137

Supelco

Etonitazene hydrochloride solution

1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Benzimidazole, 1-[2-(diethylamino)ethyl]-2-(p-ethoxybenzyl)-5-nitro-, hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N4O3 · HCl
CAS Number:
Molecular Weight:
432.94
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product E-137 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

description

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol (as free base)

format

single component solution

storage temp.

-10 to -25°C

SMILES string

[Cl-].[N+H](CC[n]1c2c(nc1Cc3ccc(cc3)OCC)cc(cc2)[N+](=O)[O-])(CC)CC

InChI

1S/C22H28N4O3.ClH/c1-4-24(5-2)13-14-25-21-12-9-18(26(27)28)16-20(21)23-22(25)15-17-7-10-19(11-8-17)29-6-3;/h7-12,16H,4-6,13-15H2,1-3H3;1H

InChI key

JTBRYBUIGVMMQL-UHFFFAOYSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service