Skip to Content
Merck
All Photos(1)

Key Documents

D-093

Supelco

3,4-Dimethylmethcathinone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO · HCl
CAS Number:
Molecular Weight:
227.73
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC(NC)C(C1=CC=C(C)C(C)=C1)=O.Cl

InChI

1S/C12H17NO.ClH/c1-8-5-6-11(7-9(8)2)12(14)10(3)13-4;/h5-7,10,13H,1-4H3;1H

InChI key

XTCDQRBKOHSIMB-UHFFFAOYSA-N

General description

3,4-Dimethylmethcathinone is a designer drug analog of the bath salt mephedrone. Also known as 3,4-DMMC, this stimulant drug is sold in many bath blends across Europe, Australia, and the US. This certified solution standard is suitable for use in bath salt testing methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service