Skip to Content
Merck
All Photos(1)

Key Documents

D-082

Supelco

Desoxypipradrol hydrochloride solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N · HCl
CAS Number:
Molecular Weight:
287.83
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

C1(C(C2CCCCN2)C3=CC=CC=C3)=CC=CC=C1.Cl

InChI

1S/C18H21N.ClH/c1-3-9-15(10-4-1)18(16-11-5-2-6-12-16)17-13-7-8-14-19-17;/h1-6,9-12,17-19H,7-8,13-14H2;1H

InChI key

NTADPDIPKMRRQV-UHFFFAOYSA-N

General description

Desoxypipradrol, also referred to as 2-DPMP or 2-diphenylmethylpiperidine, is a stimulant and norepinephrine-dopamine reuptake inhibitor (NDRI). This designer drug, structurally similar to methylphenidate, is sometimes found in certain bath salt blends such as Ivory Wave. This Certified Spiking Solution® is suitable as starting material in calibrators and controls for use in LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Loi et al.
Frontiers in pharmacology, 11, 806-806 (2020-07-17)
Stimulant drugs, including novel psychoactive substances (NPS, formerly "legal highs") have addictive potential which their users may not realize. Stimulants increase extracellular dopamine levels in the brain, including the reward and addiction pathways, through interacting with dopamine transporter (DAT). This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service