Skip to Content
Merck
All Photos(1)

Key Documents

C-251

Supelco

Cannabichromeorcin (CBCO) solution

1 mg/mL (Degassed Methanol ), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22O2
CAS Number:
Molecular Weight:
258.36
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product C-251 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (Degassed Methanol )

format

single component solution

storage temp.

-10 to -25°C

General description

This certified Snap-N-Spike® reference solution is suitable for numerous GC/MS or LC/MS applications from pharmaceutical testing and research to clinical toxicology or forensic analysis. Cannabichromeorcin (CBCO) is a cannabinoid found in cannabis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

An accurate and precise HPLC-PDA measurement of 32 cannabinoids and 2 cannflavins in hemp samples using a single extraction in less than 32 minutes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service