Skip to Content
Merck
All Photos(1)

Key Documents

LM1204

Avanti

17:0-14:1 PG

Avanti Research - A Croda Brand

Synonym(s):

1-heptadecanoyl-2-(9Z-tetradecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$328.00

$328.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$328.00

About This Item

Empirical Formula (Hill Notation):
C37H74NO10P
CAS Number:
Molecular Weight:
723.96
UNSPSC Code:
12352211
NACRES:
NA.25

$328.00


Check Cart for Availability

Request a Bulk Order

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1204-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\CCCC)=O)COC(CCCCCCCCCCCCCCCC)=O.[NH4+]

InChI key

RZHZFSXZRZZKNC-OOMNRBSDSA-N

General description

Phosphatidylglycerol (PG) is widely found in the pulmonary surfactant. It is localized to the alveoli.[1]

Application

17:0-14:1 PG or 1-heptadecanoyl-2-(9Z-tetradecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) has been used as a standard for lipid quantification by liquid chromatography mass spectrometry (LC-MS).[2] It might be used as a standard to spike the pellets in lipid extraction and for the characterization of phospholipids (PLs).[3]

Biochem/physiol Actions

Phosphatidylglycerol (PG) is known to control in born defense and inflammatory mechanism against viral infection.[1]

Packaging

2 mL Amber Glass Sealed Ampule (LM1204-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chijun Li et al.
The Journal of biological chemistry, 291(48), 25144-25153 (2016-10-21)
Phosphatidylglycerol (PG) makes up 5-20% of the phospholipids of Escherichia coli and is essential for growth in wild-type cells. PG is synthesized from the dephosphorylation of its immediate precursor, phosphatidylglycerol phosphate (PGP) whose synthase in E. coli is PgsA. Using
Tanxi Cai et al.
Journal of lipid research, 57(3), 388-397 (2016-01-07)
Phospholipids (PLs), one of the lipid categories, are not only the primary building blocks of cellular membranes, but also can be split to produce products that function as second messengers in signal transduction and play a pivotal role in numerous
Kallol Gupta et al.
Nature protocols, 13(5), 1106-1120 (2018-04-28)
With the recent success in determining membrane protein structures, further detailed understanding of the identity and function of the bound lipidome is essential. Using an approach that combines high-energy native mass spectrometry (HE-nMS) and solution-phase lipid profiling, this protocol can
Rachel Fickes et al.
Rapid communications in mass spectrometry : RCM, 30(24), 2601-2606 (2016-10-01)
Structural analogs of the bioactive lipid 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoglycerol were synthesized with a xylitol polar head group and both diacyl and diether radyl groups. Mass spectral characterization of xylitol phospholipids (PX) was carried out using collisional activation and high-resolution mass measurements of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service