Skip to Content
Merck
All Photos(1)

Key Documents

900124E

Avanti

5-OxoETE

Avanti Research - A Croda Brand

Synonym(s):

5-Oxo-6E,8Z,11Z,14Z-eicosatetraenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$510.00

$510.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$510.00

About This Item

Empirical Formula (Hill Notation):
C20H30O3
CAS Number:
Molecular Weight:
318.45
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$510.00


Estimated to ship on24 May 2025


Request a Bulk Order

Assay

>99% (TLC)

form

ethanol solution

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

pkg of (pkg of 1 X 100 ug (900124E-100UG))

concentration

100 μg/mL in ethanol

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−70°C

SMILES string

OC(CCCC(/C=C/C=C\C/C=C\C/C=C\CCCCC)=O)=O

InChI

1S/C20H30O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19(21)17-15-18-20(22)23/h6-7,9-10,12-14,16H,2-5,8,11,15,17-18H2,1H3,(H,22,23)/b7-6-,10-9-,13-12-,16-14+

InChI key

MEASLHGILYBXFO-XTDASVJISA-N

Packaging

2 mL Amber Glass Sealed Ampule (900124E-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service