Skip to Content
Merck
All Photos(1)

Key Documents

900113P

Avanti

PGE1-d9

powder

Synonym(s):

Prostaglandin E1-d9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$247.00
500 μG
$990.00

$247.00


Estimated to ship on23 March 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$247.00
500 μG
$990.00

About This Item

Empirical Formula (Hill Notation):
C20H25D9O5
CAS Number:
Molecular Weight:
363.54
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.12

$247.00


Estimated to ship on23 March 2025


Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

package of 1 × 100 μg
package of 1 × 500 μg

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1CC([C@H](CCCCCCC(O)=O)[C@H]1/C=C/[C@@H](O)CC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O

InChI key

GMVPRGQOIOIIMI-XRKMKCTBSA-N

General description

Prostaglandin E1-d9 (PGE1-d9) is a deuterated derivative of PGE1.

Biochem/physiol Actions

Prostaglandin E1 (PGE1) possesses vasodilator activities.[1]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (900113-50UG)
2 mL Amber Serum Vial with Stopper and Crimp Cap (900113-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prostaglandins (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service